Succinic anhydride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12245-36
A12245-0B
A12245-0I
A12245-30
CAAAAA12245-36EA
73.31
CAD
CAAAAA12245-36
CAAAA12245-0B
CAAAA12245-0I
CAAAA12245-30
Succinic anhydride 99%
Succinic anhydride
MDL: MFCD00005525
Beilstein: 108441
Soluble in chloroform
Succinic anhydride is used as a sizing agent and curative for epoxy resin. It acts as an additive in paper production. It is utilized in the preparation of covalently cross-linked oxidized-alginate and N-succinyl-chitosan hydrogels. It serves as an intermediate for functionalized oxide surfaces on a chip and for polyester resins. Further, it finds use in cosmetics, pharmaceuticals and in coatings.
Formula:
C₄H₄O₃ Boiling Pt: 261 °C Melting Pt: 118…121 °C Density: 1.572 Flash Pt: 157 °C (314 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005525 CAS Number: 108-30-5 EINECS: 203-570-0 UN: 3261 ADR: 8,III Merck Index: 14,8870 |
Specification Test Results
Appearance (Color) | White |
Form | Powder or crystals or crystalline powder or flakes and/or chips |
Assay (Aqueous acidbase Titration) | ≥98.5 to ≤101.5% |
Identification (FTIR) | Conforms |
Melting Point (clear melt) | 116 - 121 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...