2-Chloro-4-(trifluoromethyl)aniline 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Amino-3-chloro-α,α,α-trifluorotoluene, 2-Chloro-α,α,α-trifluoro-p-toluidine, 4-Amino-3-chlorobenzotrifluoride
A12242-14
A12242-22
A12242-06
CAAAAA12242-14EA
149.31
CAD
CAAAAA12242-14
CAAAAA12242-22
CAAAA12242-06
2-Chloro-4-(trifluoromethyl)aniline 98%
2-Chloro-4-(trifluoromethyl)aniline
Purity: 98%
CAS No.: 39885-50-2
Molecular Formula: C7H5ClF3N
Formula Weight: 195.57
Flash Point: 103°(217°F)
Refractive Index: 1.5015
Boiling Point: 214-218°
Beilstein Reference: 4179703
UN No.: 2810
MDL No.: MFCD00042563
Formula:
C₇H₅ClF₃N MW: 195.57 g/mol Boiling Pt: 89 Density: 1.434 Flash Pt: 103 °C (217 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00042563 CAS Number: 39885-50-2 EINECS: 254-674-8 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...