3,5-Xylenol 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 5-Hydroxy-3-xylene, 3,5-Dimethylphenol, 5-Hydroxy-m-xylene, 1,3-Dimethyl-5-hydroxybenzene

A12236-22 A12236-36
CAAAA12236-22EA 60.2 CAD
CAAAA12236-22 CAAAA12236-36
3,5-Xylenol 98+%
3,5-Xylenol

Purity: 98+% CAS No.: 108-68-9 Molecular Formula: C8H10O Formula Weight: 122.17 Flash Point: 109°(228°F) Melting Point: 61-65° Boiling Point: 222° Beilstein Reference: 774117 Merck Reference: 14,10082 UN No.: 2261 MDL No.: MFCD00002307

Formula: C₈H₁₀O
MW: 122.17 g/mol
Boiling Pt: 222
Melting Pt: 61…65 °C
Density: 1.12
Flash Pt: 109 °C (228 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002307
CAS Number: 108-68-9
EINECS: 203-606-5
UN: 2928
ADR: 6.1 II
Merck Index: 14,10082

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR