DL-Alanine 99%

Supplier: Thermo Scientific Chemicals

Synonyms: (±)-Alanine, DL-α-Alanine, H-DL-Ala-OH, α-Alanine

A12230-0B A12230-0I A12230-30
CAAAA12230-0BEA 142.04 CAD
CAAAA12230-0B CAAAA12230-0I CAAAA12230-30
DL-Alanine 99%
DL-Alanine

MDL: MFCD00064408 Beilstein Registry No.: 635807


Purity: 99% CAS No.: 302-72-7 Molecular Formula: C3H7NO2 Formula Weight: 89.09 Melting Point: ca 292° dec. Beilstein Reference: 635807 Merck Reference: 14,204 MDL No.: MFCD00064408

Formula: C₃H₇NO₂
MW: 89.09 g/mol
Boiling Pt: 189
Melting Pt: 79 (dec.)
Density: 1.42
Storage Temperature: Ambient
MDL Number: MFCD00064408
CAS Number: 302-72-7
EINECS: 206-126-4
Merck Index: 14,204

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR