2-Fluorobiphenyl 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12227-09
A12227-06
A12227-14
CAAAAA12227-09EA
237
CAD
CAAAAA12227-09
CAAAA12227-06
CAAAA12227-14
2-Fluorobiphenyl 98%
2-Fluorobiphenyl
MDL: MFCD00000317
Beilstein Registry No.: 2043175
Purity: 98%
CAS No.: 321-60-8
Molecular Formula: C12H9F
Formula Weight: 172.20
Melting Point: 72-76°
Boiling Point: 248°
MDL No.: MFCD00000317
Formula:
C₁₂H₉F Boiling Pt: 248 °C Melting Pt: 72…76 °C Density: 1.245 Storage Temperature: Ambient |
MDL Number:
MFCD00000317 CAS Number: 321-60-8 EINECS: 206-290-7 UN: 3077 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...