1-Hexyl isothiocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

A12223-06 A12223-14
CAAAA12223-06EA 89.27 CAD
CAAAA12223-06 CAAAA12223-14
1-Hexyl isothiocyanate 97%
1-Hexyl isothiocyanate

MDL: MFCD00014445 Beilstein Registry No.: 1751837


Purity: 97% CAS No.: 4404-45-9 Molecular Formula: CH3(CH2)5NCS Formula Weight: 143.25 Flash Point: 90°(194°F) Refractive Index: 1.4915 Odor: Pungent Boiling Point: 216° Beilstein Reference: 1751837 UN No.: 2922 MDL No.: MFCD00014445

Formula: C₇H₁₃NS
Boiling Pt: 216 °C
Density: 0.933
Flash Pt: 90 °C (194 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014445
CAS Number: 4404-45-9
EINECS: 224-549-2
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR