4'-Bromoacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

A12222-14 A12222-22 A12222-36
CAAAA12222-14EA 43.92 CAD
CAAAA12222-14 CAAAA12222-22 CAAAA12222-36
4'-Bromoacetophenone 98%
4'-Bromoacetophenone

MDL: MFCD00000105 Beilstein Registry No.: 386015


Purity: 98% CAS No.: 99-90-1 Molecular Formula: C8H7BrO Formula Weight: 199.05 Flash Point: 110°(230°F) Melting Point: 49-53° Boiling Point: 255-256° Beilstein Reference: 386015 Merck Reference: 14,1403 MDL No.: MFCD00000105

Formula: C₈H₇BrO
Boiling Pt: 255…256 °C
Melting Pt: 49…53 °C
Density: 1.736 g/cm³ (20 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000105
CAS Number: 99-90-1
EINECS: 202-799-3
UN: 2811
ADR: 6.1,II
Merck Index: 14,1403

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR