Tyramine hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(4-Hydroxyphenyl)ethylammonium chloride, 4-(2-Aminoethyl)phenol hydrochloride, 4-Hydroxyphenethylammonium chloride
A12220-06
A12220-14
A12220-22
CAAAA12220-06EA
46.14
CAD
CAAAA12220-06
CAAAA12220-14
CAAAA12220-22
Tyramine hydrochloride 98%
Tyramine hydrochloride
Reagent for fluorescence enzyme immunoassay
Purity: 98%
CAS No.: 60-19-5
Molecular Formula: C8H11NO.HCl
Formula Weight: 173.65
Melting Point: 271-274°
Beilstein Reference: 3627058
Merck Reference: 14,9835
MDL No.: MFCD00012901
Formula:
C₈H₁₂ClNO MW: 173.64 Melting Pt: 271…274 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012901 CAS Number: 60-19-5 EINECS: 200-462-5 Merck Index: 14,9835 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...