5-Bromo-2-chlorobenzoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A12218-09 A12218-18 A12218-30
CAAAA12218-09EA 49.61 CAD
CAAAA12218-09 CAAAA12218-18 CAAAA12218-30
5-Bromo-2-chlorobenzoic acid 98+%
5-Bromo-2-chlorobenzoic acid

MDL: MFCD00002415 Beilstein Registry No.: 2691432


Purity: 98+% CAS No.: 21739-92-4 Molecular Formula: C7H4BrClO2 Formula Weight: 235.47 Odor: Odorless Melting Point: 157-162° Beilstein Reference: 2691432 MDL No.: MFCD00002415

Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
Boiling Pt: 316 °C (1013 hPa)
Melting Pt: 157…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00002415
CAS Number: 21739-92-4
EINECS: 244-558-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR