3,5-Dimethylisoxazole 99%

Supplier: Thermo Scientific Chemicals
Warning

A12217-14 A12217-22
CAAAA12217-14EA 77.26 CAD
CAAAA12217-14 CAAAA12217-22
3,5-Dimethylisoxazole 99%
3,5-Dimethylisoxazole

MDL: MFCD00003156 Beilstein Registry No.: 106324 Fieser: 18,205


Purity: 99% CAS No.: 300-87-8 Molecular Formula: C5H7NO Formula Weight: 97.12 Flash Point: 31°(87°F) Refractive Index: 1.4415 Melting Point: -14° Boiling Point: 142-144° Beilstein Reference: 106324 UN No.: 1993 MDL No.: MFCD00003156

Formula: C₅H₇NO
MW: 97.12 g/mol
Boiling Pt: 142…144 °C
Melting Pt: –14 °C
Density: 0.992
Flash Pt: 31 °C (87 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003156
CAS Number: 300-87-8
EINECS: 206-100-2
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR