3'-(Trifluoromethyl)acetophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12210-06
A12210-14
CAAAAA12210-03EA
39.48
CAD
CAAAAA12210-03
CAAAA12210-06
CAAAA12210-14
3'-(Trifluoromethyl)acetophenone 98+%
3'-(Trifluoromethyl)acetophenone
MDL: MFCD00000391
Beilstein Registry No.: 640151
Formula:
C₉H₇F₃O Boiling Pt: 198…200 °C Density: 1,260 Flash Pt: 83 °C(181 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000391 CAS Number: 349-76-8 EINECS: 206-490-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...