2,6-Dimethoxybenzaldehyde 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12209-03
A12209-06
A12209-14
CAAAA12209-03EA
80.74
CAD
CAAAA12209-03
CAAAA12209-06
CAAAA12209-14
2,6-Dimethoxybenzaldehyde 98+%
2,6-Dimethoxybenzaldehyde
Purity: 98+%
CAS No.: 3392-97-0
Molecular Formula: C9H10O3
Formula Weight: 166.18
Melting Point: 96-98°
Boiling Point: 285°
MDL No.: MFCD00010862
Formula:
C₉H₁₀O₃ Boiling Pt: 285 °C Melting Pt: 96…98 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010862 CAS Number: 3392-97-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...