2-Chlorobenzimidazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12193-06
A12193-14
CAAAA12193-06EA
127.19
CAD
CAAAA12193-06
CAAAA12193-14
CAAAA12193-22
2-Chlorobenzimidazole 97%
2-Chlorobenzimidazole
MDL: MFCD00051944
Beilstein Registry No.: 116526
Purity: 97%
CAS No.: 4857-06-1
Molecular Formula: C7H5ClN2
Formula Weight: 152.58
Melting Point: ca 180-185°
Beilstein Reference: 116526
Formula:
C₇H₅ClN₂ Melting Pt: ca 180…185 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051944 CAS Number: 4857-06-1 EINECS: 225-453-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...