Benzyl(dimethyl)amine 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
DMBA, N,N-Dimethylbenzylamine, N-Benzyldimethylamine, BDMA
A12191-0J
A12191-AK
A12191-AU
CAAAA12191-0JEA
709.42
CAD
CAAAA12191-0J
CAAAA12191-AK
CAAAA12191-AU
Benzyl(dimethyl)amine 98+%
Benzyl(dimethyl)amine
MDL: MFCD00008329
Beilstein Registry No.: 1099620
Purity: 98+%
CAS No.: 103-83-3
Molecular Formula: C9H13N
Formula Weight: 135.21
Flash Point: 54°(129°F)
Refractive Index: 1.5010
Melting Point: -75°
Boiling Point: 183-184°
Beilstein Reference: 1099620
UN No.: 2619
MDL No.: MFCD00008329
Formula:
C₆H₅CH₂N(CH₃)₂ MW: 135.21 g/mol Boiling Pt: 183 Melting Pt: -75 Density: 0.900 Flash Pt: 54 °C (129 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008329 CAS Number: 103-83-3 EINECS: 203-149-1 UN: 2619 ADR: 8 II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...