α,α,α-Trifluoro-4-iodotoluene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Iodobenzotrifluoride, 1-Iodo-4-(trifluoromethyl)benzene, α,α,α-Trifluoro-p-iodotoluene
A12189-03
A12189-06
A12189-14
CAAAA12189-03EA
41.08
CAD
CAAAA12189-03
CAAAA12189-06
CAAAA12189-14
α,α,α-Trifluoro-4-iodotoluene 98%
α,α,α-Trifluoro-4-iodotoluene
MDL: MFCD00039398
Beilstein Registry No.: 1944013
Purity: 98%
CAS No.: 455-13-0
Molecular Formula: C7H4F3I
Formula Weight: 272.01
Flash Point: 68°(154°F)
Refractive Index: 1.5180
Melting Point: -8°
Boiling Point: 68-70°/15mm
Beilstein Reference: 1944013
MDL No.: MFCD00039398
Formula:
C₇H₄F₃I MW: 272.01 g/mol Boiling Pt: 68…70 °C (15 mmHg) Melting Pt: 17 Density: 1.851 Flash Pt: 68 °C (154 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039398 CAS Number: 455-13-0 EINECS: 207-234-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...