3',4'-Dimethylacetophenone 98%

Supplier: Thermo Scientific Chemicals
Warning

A12187-06 A12187-14 A12187-22
CAAAA12187-06EA 40.45 CAD
CAAAA12187-06 CAAAA12187-14 CAAAA12187-22
3',4'-Dimethylacetophenone 98%
3',4'-Dimethylacetophenone

MDL: MFCD00008743 Beilstein Registry No.: 907053


Purity: 98% CAS No.: 3637-01-2 Molecular Formula: C10H12O Formula Weight: 148.21 Flash Point: 102°(215°F) Refractive Index: 1.5380 Boiling Point: 242-243° Beilstein Reference: 907053 MDL No.: MFCD00008743

Formula: C₁₀H₁₂O
Boiling Pt: 242…243 °C
Melting Pt: –3 °C
Density: 1.002
Flash Pt: 102 °C(215 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008743
CAS Number: 3637-01-2
EINECS: 222-859-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR