2',5'-Dihydroxyacetophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(2,5-Dihydroxyphenyl)ethan-1-one, 1-(2,5-Dihydroxyphenyl)ethanone, 2,5-Dihydroxyacetophenone
A12185-06
A12185-14
A12185-22
CAAAA12185-06EA
65.25
CAD
CAAAA12185-06
CAAAA12185-14
CAAAA12185-22
2',5'-Dihydroxyacetophenone 98+%
2',5'-Dihydroxyacetophenone
MDL: MFCD00002343
Beilstein Registry No.: 637903
Purity: 98+%
CAS No.: 490-78-8
Molecular Formula: C8H8O3
Formula Weight: 152.15
Melting Point: 204-208°
Beilstein Reference: 637903
Formula:
C₈H₈O₃ MW: 152.15 g/mol Boiling Pt: 296 °C (1013 hPa) Melting Pt: 204…208 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002343 CAS Number: 490-78-8 EINECS: 207-716-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...