Sudan IV
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12181-14
A12181-22
CAAAA12181-14EA
58.14
CAD
CAAAA12181-14
CAAAA12181-22
Sudan IV
Sudan IV
Useful as a stain for fat in animal tissue
CAS No.: 85-83-6
Molecular Formula: C24H20N4O
Formula Weight: 380.45
Melting Point: ca 185° dec.
Merck Reference: 14,8393
MDL No.: MFCD00003893
Formula:
C₂₄H₂₀N₄O Melting Pt: ∼ 185 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00003893 CAS Number: 85-83-6 EINECS: 201-635-8 Merck Index: 14,8393 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...