N-(2-Bromoethyl)phthalimide 98+%
Supplier: Thermo Scientific Chemicals
A12164-14
A12164-22
A12164-36
CAAAA12164-14EA
70.78
CAD
CAAAA12164-14
CAAAA12164-22
CAAAA12164-36
N-(2-Bromoethyl)phthalimide 98+%
N-(2-Bromoethyl)phthalimide
MDL: MFCD00005902
Beilstein Registry No.: 148736
Purity: 98+%
CAS No.: 574-98-1
Molecular Formula: C10H8BrNO2
Formula Weight: 254.09
Melting Point: 80-84°
Beilstein Reference: 148736
Formula:
C₁₀H₈BrNO₂ Boiling Pt: 318 °C (1013 hPa) Melting Pt: 80…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005902 CAS Number: 574-98-1 EINECS: 209-379-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...