2-Chloro-3-nitropyridine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12161-22
A12161-06
A12161-14
CAAAAA12161-22EA
445.56
CAD
CAAAAA12161-22
CAAAA12161-06
CAAAA12161-14
2-Chloro-3-nitropyridine 99%
2-Chloro-3-nitropyridine
MDL: MFCD00006232
Beilstein Registry No.: 124482
Purity: 99%
CAS No.: 5470-18-8
Molecular Formula: C5H3ClN2O2
Formula Weight: 158.54
Flash Point: 185°(365°F)
Melting Point: 99-103°
Beilstein Reference: 124482
MDL No.: MFCD00006232
Formula:
C₅H₃ClN₂O₂ MW: 158.54 g/mol Boiling Pt: 260 °C (1013 hPa) Melting Pt: 99…103 °C Flash Pt: 185 °C(365 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006232 CAS Number: 5470-18-8 EINECS: 226-799-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...