Methyl violet 2B

Supplier: Thermo Scientific Chemicals
Warning

A12159-18 A12159-30
CAAAA12159-18EA 67.47 CAD
CAAAA12159-18 CAAAA12159-30
Methyl violet 2B
Methyl Violet 2B

MDL: MFCD00012568 Notes: Transition interval: pH 0.0 (yellow) to pH 1.6 (blue)


CAS No.: 8004-87-3 Molecular Formula: C24H27N3.HCl Formula Weight: 393.96 Odor: Odorless MDL No.: MFCD00012568

Formula: C₂₄H₂₈ClN₃
Melting Pt: 137 °C
Storage Temperature: Ambient
MDL Number: MFCD00012568
CAS Number: 8004-87-3
UN: UN3077
ADR: 9,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR