4-Methylmorpholine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12158-0F
A12158-AE
A12158-AP
CAAAA12158-0FEA
282.82
CAD
CAAAA12158-0F
CAAAA12158-AE
CAAAA12158-AP
4-Methylmorpholine 99%
4-Methylmorpholine
MDL: MFCD00006175
Beilstein Registry No.: 102719
Fieser: 1,690 2,278
Purity: 99%
CAS No.: 109-02-4
Molecular Formula: C5H11NO
Formula Weight: 101.15
Flash Point: 14°(57°F)
Physical Form: Liquid
Density: 0.920
Refractive Index: 1.4355
Appearance: Colorless
Odor: Amine-like
pH: 10.6
Melting Point: -65°
Boiling Point: 116-118°
Beilstein Reference: 102719
Merck Reference: 14,6277
UN No.: 2535
MDL No.: MFCD00006175
Formula:
C₅H₁₁NO Boiling Pt: 116…118 °C Melting Pt: –65 °C Density: 0.920 Flash Pt: 14 °C (57 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00006175 CAS Number: 109-02-4 EINECS: 203-640-0 UN: 2535 ADR: 3,II Merck Index: 14,6277 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...