Toluene-4-sulfonamide 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: p-Toluenesulfonamide, 4-Methylbenzenesulfonamide, PTSA, 4-Toluenesulfonamide, p-Toluenesulphonamide, 4-Methylbenzenesulphonamide, 4-Toluenesulphonamide

A12156-30 A12156-0C A12156-0E A12156-36
CAAAAA12156-30EA 45.98 CAD
CAAAAA12156-30 CAAAA12156-0C CAAAA12156-0E CAAAA12156-36
Toluene-4-sulfonamide 98+%
Toluene-4-sulphonamide

MDL: MFCD00011692 Beilstein Registry No.: 472689


Purity: 98+% CAS No.: 70-55-3 Molecular Formula: C7H9NO2S Formula Weight: 171.22 Flash Point: 202°(395°F) Melting Point: 136-139° Boiling Point: 221°/10mm Beilstein Reference: 472689 MDL No.: MFCD00011692

Formula: C₇H₉NO₂S
MW: 171.22 g/mol
Boiling Pt: 221 °C/10 mm
Melting Pt: 136…139 °C
Flash Pt: 202 °C(395 °F)
Storage Temperature: Ambient
MDL Number: MFCD00011692
CAS Number: 70-55-3
EINECS: 200-741-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR