3'-Nitroacetophenone 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: m-Nitroacetophenone

A12155-36 A12155-0B A12155-30
CAAAAA12155-36EA 224.36 CAD
CAAAAA12155-36 CAAAA12155-0B CAAAA12155-30
3'-Nitroacetophenone 98+%
3'-Nitroacetophenone

MDL: MFCD00007259 Beilstein Registry No.: 743002


Purity: 98+% CAS No.: 121-89-1 Molecular Formula: C8H7NO3 Formula Weight: 165.15 Odor: Slightly sweetish Melting Point: 76-80° Boiling Point: 202° Beilstein Reference: 743002 MDL No.: MFCD00007259

Formula: C₈H₇NO₃
MW: 165.15 g/mol
Boiling Pt: 202 °C
Melting Pt: 76…80 °C
Density: 1.433 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007259
CAS Number: 121-89-1
EINECS: 204-504-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR