4-tert-Butylthiophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-tert-Butyl-4-mercaptobenzene, 4-tert-Butylbenzene-1-thiol, 4-tert-Butylbenzenethiol

A12148-03 A12148-06 A12148-14
CAAAA12148-03EA 43.61 CAD
CAAAA12148-03 CAAAA12148-06 CAAAA12148-14
4-tert-Butylthiophenol 97%
4-tert-Butylthiophenol

MDL: MFCD00022067 Beilstein Registry No.: 606476


Purity: 97% CAS No.: 2396-68-1 Molecular Formula: C10H14S Formula Weight: 166.29 Flash Point: 110°(230°F) Refractive Index: 1.5485 Odor: Stench Melting Point: -11° Boiling Point: 238° Beilstein Reference: 606476 MDL No.: MFCD00022067

Formula: C₁₀H₁₄S
MW: 166.29 g/mol
Boiling Pt: 238 °C
Melting Pt: –11 °C
Density: 0.990
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00022067
CAS Number: 2396-68-1
EINECS: 219-255-6
UN: 3334
ADR: 9,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR