4-Iodoanisole 98+%

Supplier: Thermo Scientific Chemicals

A12144-14 A12144-22 A12144-36
CAAAA12144-14EA 57.04 CAD
CAAAA12144-14 CAAAA12144-22 CAAAA12144-36
4-Iodoanisole 98+%
4-Iodoanisole

MDL: MFCD00001056 Beilstein Registry No.: 1906692


Purity: 98+% CAS No.: 696-62-8 Molecular Formula: C7H7IO Formula Weight: 234.04 Flash Point: 110°(230°F) Melting Point: 48-52° Boiling Point: 240° Beilstein Reference: 1906692 MDL No.: MFCD00001056

Formula: C₇H₇IO
MW: 234.04 g/mol
Boiling Pt: 240 °C
Melting Pt: 48…52 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001056
CAS Number: 696-62-8
EINECS: 211-798-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR