Phenylacetylene 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12139-14
A12139-22
A12139-36
CAAAA12139-14EA
73.31
CAD
CAAAA12139-14
CAAAA12139-22
CAAAA12139-36
Phenylacetylene 98+%
Phenylacetylene
MDL: MFCD00008570
Beilstein Registry No.: 605461
Purity: 98+%
CAS No.: 536-74-3
Molecular Formula: C8H6
Formula Weight: 102.14
Flash Point: 27°(81°F)
Refractive Index: 1.5480
Melting Point: -45°
Boiling Point: 142-144°
Beilstein Reference: 605461
Merck Reference: 14,3861
UN No.: 3295
MDL No.: MFCD00008570
Formula:
C₈H₆ MW: 102.14 g/mol Boiling Pt: 142…144 °C Melting Pt: –45 °C Density: 0.930 Flash Pt: 27 °C (81 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008570 CAS Number: 536-74-3 EINECS: 208-645-1 UN: 3295 ADR: 3 III Merck Index: 14,3861 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...