Octylbenzene 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-Phenyloctane
A12138-14
A12138-22
A12138-36
CAAAA12138-14EA
170.64
CAD
CAAAA12138-14
CAAAA12138-22
CAAAA12138-36
Octylbenzene 99%
Octylbenzene
MDL: MFCD00009564
Beilstein Registry No.: 1906253
Purity: 99%
CAS No.: 2189-60-8
Molecular Formula: C14H22
Formula Weight: 190.33
Flash Point: 107°(224°F)
Refractive Index: 1.4840
Odor: Mild
Melting Point: -36°
Boiling Point: 264-265°
Beilstein Reference: 1906253
MDL No.: MFCD00009564
Formula:
C₁₄H₂₂ MW: 190.33 g/mol Boiling Pt: 264…265 °C Melting Pt: –36 °C Density: 0.854 Flash Pt: 107 °C(224 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009564 CAS Number: 2189-60-8 EINECS: 218-582-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...