2,6-Dinitroaniline 98%

Supplier: Thermo Scientific Chemicals
Danger

A12120-06 A12120-14 A12120-22
CAAAA12120-06EA 91.64 CAD
CAAAA12120-06 CAAAA12120-14 CAAAA12120-22
2,6-Dinitroaniline 98%
2,6-Dinitroaniline

MDL: MFCD00007148 Beilstein Registry No.: 2214886


Purity: 98% CAS No.: 606-22-4 Molecular Formula: C6H5N3O4 Formula Weight: 183.12 Melting Point: ca 136° dec. Beilstein Reference: 2214886 Merck Reference: 14,3271 UN No.: 1596 MDL No.: MFCD00007148

Formula: C₆H₅N₃O₄
Melting Pt: ∼ 136 °C (dec.)
Density: 1.629 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007148
CAS Number: 606-22-4
EINECS: 210-108-1
UN: 1596
ADR: 6.1,II
Merck Index: 14,3271

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR