2,3,4,5,6-Pentafluorobenzyl alcohol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12119-06
A12119-14
CAAAA12119-06EA
87.85
CAD
CAAAA12119-06
CAAAA12119-14
2,3,4,5,6-Pentafluorobenzyl alcohol 98%
2,3,4,5,6-Pentafluorobenzyl alcohol
MDL: MFCD00004602
Beilstein Registry No.: 2052669
Purity: 98%
CAS No.: 440-60-8
Molecular Formula: C7H3F5O
Formula Weight: 198.09
Flash Point: 87°(188°F)
Melting Point: 32-36°
Boiling Point: 182°
Beilstein Reference: 2052669
MDL No.: MFCD00004602
Formula:
C₇H₃F₅O Boiling Pt: 182 °C Melting Pt: 32…36 °C Flash Pt: 87 °C(188 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004602 CAS Number: 440-60-8 EINECS: 207-126-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...