4'-Chloroacetophenone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12115-0B
A12115-30
CAAAA12115-0BEA
325.48
CAD
CAAAA12115-0B
CAAAA12115-0I
CAAAA12115-30
4'-Chloroacetophenone 98+%
4'-Chloroacetophenone
MDL: MFCD00000624
Beilstein Registry No.: 386014
Purity: 98+%
CAS No.: 99-91-2
Molecular Formula: C8H7ClO
Formula Weight: 154.60
Flash Point: 90°(194°F)
Refractive Index: 1.5550
Melting Point: 17-20°
Boiling Point: 230-232°
Beilstein Reference: 386014
Merck Reference: 14,2116
UN No.: 3416
MDL No.: MFCD00000624
Formula:
C₈H₇ClO Boiling Pt: 230…232 °C Melting Pt: 17…20 °C Density: 1.190 Flash Pt: 90 °C(194 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000624 CAS Number: 99-91-2 EINECS: 202-800-7 UN: 3416 ADR: 6.1,II Merck Index: 14,2116 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...