(3-Fluoro-4-hydroxyphenyl)acetic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

A12114-03 A12114-06
CAAAA12114-03EA 46.45 CAD
CAAAA12114-03 CAAAA12114-06
(3-Fluoro-4-hydroxyphenyl)acetic acid 98%
(3-Fluoro-4-hydroxyphenyl)acetic acid

MDL: MFCD00004348


Purity: 98% CAS No.: 458-09-3 Molecular Formula: C8H7FO3 Formula Weight: 170.14 Melting Point: 130-133° MDL No.: MFCD00004348

Formula: C₈H₇FO₃
Melting Pt: 130…133 °C
Storage Temperature: Ambient
MDL Number: MFCD00004348
CAS Number: 458-09-3
EINECS: 207-275-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR