N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12107-09
A12107-18
A12107-30
CAAAA12107-09EA
121.91
CAD
CAAAA12107-09
CAAAA12107-18
CAAAA12107-30
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride 98+%
N,N,N',N'-Tetramethyl-p-phenylenediamine dihydrochloride
MDL: MFCD00012482
Beilstein Registry No.: 5132096
Purity: 98+%
CAS No.: 637-01-4
Molecular Formula: C10H16N2.2HCl
Formula Weight: 237.17
Melting Point: ca 228° dec.
Beilstein Reference: 5132096
Formula:
C₁₀H₁₈Cl₂N₂ Melting Pt: ∼ 228 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00012482 CAS Number: 637-01-4 EINECS: 211-274-8 Merck Index: 12,09367 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...