Isobutyraldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12106-0F
A12106-AP
CAAAA12106-0FEA
129.56
CAD
CAAAA12106-0F
CAAAA12106-AP
Isobutyraldehyde 98%
Isobutyraldehyde
MDL: MFCD00006980
Beilstein Registry No.: 605330
Purity: 98%
CAS No.: 78-84-2
Molecular Formula: C4H8O
Formula Weight: 72.11
Storage Temperature: 2°C to 8°C
Flash Point: -24°(-11°F)
Physical Form: Liquid
Density: 0.794
Refractive Index: 1.3723
Sensitivity: Air Sensitive
Appearance: Colorless
Odor: Pungent
Melting Point: -65°
Boiling Point: 63-65°
Beilstein Reference: 605330
Merck Reference: 14,5154
UN No.: 2045
MDL No.: MFCD00006980
Formula:
C₄H₈O Boiling Pt: 63…65 °C Melting Pt: –65 °C Density: 0.794 Flash Pt: –24 °C (–11 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006980 CAS Number: 78-84-2 EINECS: 201-149-6 UN: 2045 ADR: 3,II Merck Index: 14,5154 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...