1-Bromo-4-(trifluoromethoxy)benzene 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(Trifluoromethoxy)bromobenzene
A12104-06
A12104-14
CAAAA12104-03EA
46.03
CAD
CAAAA12104-03
CAAAA12104-06
CAAAA12104-14
1-Bromo-4-(trifluoromethoxy)benzene 98%
1-Bromo-4-(trifluoromethoxy)benzene
MDL: MFCD00040834
Beilstein Registry No.: 2046332
Purity: 98%
CAS No.: 407-14-7
Molecular Formula: C7H4BrF3O
Formula Weight: 241.01
Flash Point: 67°(152°F)
Refractive Index: 1.4610
Odor: Unpleasant
Boiling Point: 155-157°
Beilstein Reference: 2046332
MDL No.: MFCD00040834
Formula:
C₇H₄BrF₃O MW: 241.01 g/mol Boiling Pt: 155…157 °C Density: 1.622 Flash Pt: 67 °C(152 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00040834 CAS Number: 407-14-7 EINECS: 206-979-2 UN: 3334 ADR: 9, |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...