1-Iodo-2-nitrobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

A12100-14 A12100-22
CAAAA12100-14EA 106.33 CAD
CAAAA12100-14 CAAAA12100-22
1-Iodo-2-nitrobenzene 97%
1-Iodo-2-nitrobenzene

MDL: MFCD00007088 Beilstein Registry No.: 2044110


Purity: 97% CAS No.: 609-73-4 Molecular Formula: C6H4INO2 Formula Weight: 249.01 Flash Point: 110°(230°F) Melting Point: 48-52° Boiling Point: 288-289° Beilstein Reference: 2044110 MDL No.: MFCD00007088

Formula: C₆H₄INO₂
Boiling Pt: 288…289 °C
Melting Pt: 48…52 °C
Density: 1,9186 g/cm³ (75 °C)
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007088
CAS Number: 609-73-4
EINECS: 210-200-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR