Tris(2-methylphenyl)phosphine 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12093-03
A12093-06
A12093-14
CAAAA12093-03EA
43.77
CAD
CAAAA12093-03
CAAAA12093-06
CAAAA12093-14
Tris(2-methylphenyl)phosphine 98+%
Tris(2-methylphenyl)phosphine
MDL: MFCD00008514
Beilstein Registry No.: 661212
Purity: 98+%
CAS No.: 6163-58-2
Molecular Formula: C21H21P
Formula Weight: 304.37
Melting Point: 122-127°
Beilstein Reference: 661212
Formula:
C₂₁H₂₁P Boiling Pt: 187 - 192 °C (0.05 -rr) Melting Pt: 122…127 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008514 CAS Number: 6163-58-2 EINECS: 228-193-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...