(±)-1-Octen-3-ol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12079-09
A12079-18
A12079-30
CAAAA12079-09EA
56.56
CAD
CAAAA12079-09
CAAAA12079-18
CAAAA12079-30
(±)-1-Octen-3-ol 98%
(±)-1-Octen-3-ol
MDL: MFCD00004589
Beilstein Registry No.: 1744110
Purity: 98%
CAS No.: 3391-86-4
Molecular Formula: C8H16O
Formula Weight: 128.22
Flash Point: 61°(141°F)
Refractive Index: 1.4370
Odor: Characteristic
Melting Point: -49°
Boiling Point: 173-175°
Beilstein Reference: 1744110
MDL No.: MFCD00004589
Formula:
C₈H₁₆O Boiling Pt: 173…175 °C Melting Pt: –49 °C Density: 0.833 Flash Pt: 61 °C(141 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00004589 CAS Number: 3391-86-4 EINECS: 222-226-0 UN: 2810 ADR: 6.1,III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...