4-Butoxyphenol 98%

Supplier: Thermo Scientific Chemicals
Warning

A12076-06 A12076-14 A12076-22
CAAAA12076-06EA 92.9 CAD
CAAAA12076-06 CAAAA12076-14 CAAAA12076-22
4-Butoxyphenol 98%
4-Butoxyphenol

MDL: MFCD00002336


Purity: 98% CAS No.: 122-94-1 Molecular Formula: C10H14O2 Formula Weight: 166.22 Melting Point: 63-66° MDL No.: MFCD00002336

Formula: C₁₀H₁₄O₂
Boiling Pt: 125 °C (4 -rr)
Melting Pt: 63…66 °C
Storage Temperature: Ambient
MDL Number: MFCD00002336
CAS Number: 122-94-1
EINECS: 204-583-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR