Vanillic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Hydroxy-3-methoxybenzoic acid

A12074-14 A12074-22 A12074-36
CAAAA12074-14EA 68.89 CAD
CAAAA12074-14 CAAAA12074-22 CAAAA12074-36
Vanillic acid 98%
Vanillic acid

MDL: MFCD00002551 Beilstein Registry No.: 2208364


Purity: 98% CAS No.: 121-34-6 Molecular Formula: C8H8O4 Formula Weight: 168.15 Melting Point: 210-218° Beilstein Reference: 2208364 Merck Reference: 14,9931 MDL No.: MFCD00002551

Formula: C₈H₈O₄
MW: 168.15 g/mol
Boiling Pt: 353
Melting Pt: 210…218 °C
Storage Temperature: Ambient
MDL Number: MFCD00002551
CAS Number: 121-34-6
EINECS: 204-466-8
Merck Index: 14,9931

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR