Myristic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Tetradecylic acid, Tetradecanoic acid

A12067-30 A12067-0E A12067-36
CAAAAA12067-30EA 47.08 CAD
CAAAAA12067-30 CAAAA12067-0E CAAAA12067-36
Myristic acid 98%
Myristic acid

MDL: MFCD00002744 Beilstein Registry No.: 508624


Purity: 98% CAS No.: 544-63-8 Molecular Formula: CH3(CH2)12CO2H Formula Weight: 228.38 Flash Point: 175°(347°F) Melting Point: 53-56° Boiling Point: 326° Beilstein Reference: 508624 Merck Reference: 14,6333 MDL No.: MFCD00002744

Formula: C₁₄H₂₈O₂
MW: 228.38 g/mol
Boiling Pt: 319
Melting Pt: 53…56 °C
Density: 0.862
Flash Pt: 175 °C (347 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002744
CAS Number: 544-63-8
EINECS: 208-875-2
Merck Index: 14,6333

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR