2-Amino-5-chloropyridine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chloropyridin-2-amine, 5-Chloro-2-pyridylamine

A12063-14 A12063-22 A12063-36
CAAAA12063-14EA 78.68 CAD
CAAAA12063-14 CAAAA12063-22 CAAAA12063-36
2-Amino-5-chloropyridine 98%
2-Amino-5-chloropyridine

MDL: MFCD00006324 Beilstein Registry No.: 108735


Purity: 98% CAS No.: 1072-98-6 Molecular Formula: C5H5ClN2 Formula Weight: 128.56 Flash Point: 160°(320°F) Melting Point: 135-138° Boiling Point: 127-129°/21mm Beilstein Reference: 108735 MDL No.: MFCD00006324

Formula: C₅H₅ClN₂
MW: 128.56 g/mol
Boiling Pt: 241
Melting Pt: 135…138 °C
Flash Pt: 160 °C (320 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006324
CAS Number: 1072-98-6
EINECS: 214-020-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR