4,4-Dimethoxybutanone tech. 90%, tech.
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12058-22
A12058-0B
A12058-30
CAAAAA12058-22EA
87.69
CAD
CAAAAA12058-22
CAAAA12058-0B
CAAAA12058-30
4,4-Dimethoxybutanone tech. 90%, tech.
4,4-Dimethoxybutanone
MDL: MFCD00008789
Beilstein Registry No.: 1702372
Purity: 90%
CAS No.: 5436-21-5
Molecular Formula: C6H12O3
Formula Weight: 132.16
Flash Point: 57°(134°F)
Refractive Index: 1.4150
Melting Point: -83°
Boiling Point: 170-172°
Beilstein Reference: 1702372
UN No.: 1224
MDL No.: MFCD00008789
Formula:
C₆H₁₂O₃ Boiling Pt: 170…172 °C Melting Pt: –83 °C Density: 0.996 Flash Pt: 57 °C (134 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008789 CAS Number: 5436-21-5 EINECS: 226-605-1 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...