2-Bromo-1,4-dichlorobenzene 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromo-2,5-dichlorobenzene

A12056-06 A12056-14
CAAAA12056-06EA 67.31 CAD
CAAAA12056-06 CAAAA12056-14
2-Bromo-1,4-dichlorobenzene 98%
2-Bromo-1,4-dichlorobenzene

MDL: MFCD00018505 Beilstein Registry No.: 1934815


Purity: 98% CAS No.: 1435-50-3 Molecular Formula: C6H3BrCl2 Formula Weight: 225.91 Flash Point: 110°(230°F) Melting Point: 32-36° Boiling Point: 118-120°/20mm Beilstein Reference: 1934815 MDL No.: MFCD00018505

Formula: C₆H₃BrCl₂
MW: 225.9 g/mol
Boiling Pt: 118…120 °C/20 mm
Melting Pt: 32…36 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00018505
CAS Number: 1435-50-3
EINECS: 215-859-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR