2-Phenylbenzoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Biphenyl-2-carboxylic acid, 2-Biphenylcarboxylic acid, [1,1'-Biphenyl]-2-carboxylic acid
A12049-22
A12049-06
A12049-14
CAAAAA12049-22EA
347.6
CAD
CAAAAA12049-22
CAAAA12049-06
CAAAA12049-14
2-Phenylbenzoic acid 98%
2-Phenylbenzoic acid
MDL: MFCD00002463
Beilstein Registry No.: 974075
Purity: 98%
CAS No.: 947-84-2
Molecular Formula: C13H10O2
Formula Weight: 198.22
Odor: Odorless
Melting Point: 110-114°
Boiling Point: 343-344°
Beilstein Reference: 974075
MDL No.: MFCD00002463
Formula:
C₁₃H₁₀O₂ MW: 198.22 g/mol Boiling Pt: 343…344 °C Melting Pt: 110…114 °C Density: 1.458 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002463 CAS Number: 947-84-2 EINECS: 213-432-1 |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...