Pyridoxine hydrochloride 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Adermine hydrochloride, Vitamin B6 hydrochloride, PN HCl, Pyridoxine HCl, (5-Hydroxy-6-methylpyridine-3,4-diyl)dimethanol hydrochloride, Pyridoxol hydrochloride, Pyridoxine HCI

A12041-14 A12041-22 A12041-36
CAAAA12041-14EA 82.44 CAD
CAAAA12041-14 CAAAA12041-22 CAAAA12041-36
Pyridoxine hydrochloride 99%
Pyridoxine hydrochloride

MDL: MFCD00012807 Beilstein Registry No.: 3632435


Purity: 99% CAS No.: 58-56-0 Molecular Formula: C8H11NO3.HCl Formula Weight: 205.64 Odor: Odorless pH: 3 Melting Point: ca 206° dec. Beilstein Reference: 3632435 Merck Reference: 14,7982 MDL No.: MFCD00012807

Formula: C₈H₁₂ClNO₃
MW: 205.64 g/mol
Melting Pt: ca 206 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012807
CAS Number: 58-56-0
EINECS: 200-386-2
Merck Index: 14,7982

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR