N,O-Bis(trimethylsilyl)trifluoroacetamide 98+%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Trimethylsilyl 2,2,2-trifluoro-N-(trimethylsilyl)acetimidate, Bis(Trimethylsilyl)Trifluoroacetamide, BSTFA
A12033-09
A12033-18
CAAAAA12033-22EA
681.72
CAD
CAAAAA12033-22
CAAAA12033-09
CAAAA12033-18
N,O-Bis(trimethylsilyl)trifluoroacetamide 98+%
N,O-Bis(trimethylsilyl)trifluoroacetamide
MDL: MFCD00008269
Beilstein Registry No.: 3607620
Purity: 98+%
CAS No.: 25561-30-2
Molecular Formula: C8H18F3NOSi2
Formula Weight: 257.40
Storage Temperature: 2°C to 8°C
Flash Point: 23°(73°F)
Refractive Index: 1.3840
Melting Point: -10°
Boiling Point: 145-147°
Beilstein Reference: 3607620
UN No.: 2924
MDL No.: MFCD00008269
Formula:
C₈H₁₈F₃NOSi₂ MW: 257.4 g/mol Boiling Pt: 145…147 °C Melting Pt: –10 °C Density: 0.97 Flash Pt: 23 °C (73 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00008269 CAS Number: 25561-30-2 EINECS: 247-103-9 UN: 2924 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...