2-Aminothiazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Thiazol-2-ylamine, Aminothiazole, Thiazol-2-amine
A12026-18
A12026-30
A12026-0E
A12026-22
A12026-36
CAAAAA12026-18EA
38.55
CAD
CAAAAA12026-18
CAAAAA12026-30
CAAAA12026-0E
CAAAA12026-22
CAAAA12026-36
2-Aminothiazole 97%
2-Aminothiazole
MDL: MFCD00005325
Beilstein Registry No.: 105738
Fieser: 4,19
Purity: 97%
CAS No.: 96-50-4
Molecular Formula: C3H4N2S
Formula Weight: 100.14
Melting Point: 88-92°
Boiling Point: 117°/15mm
Merck Reference: 14,479
MDL No.: MFCD00005325
Formula:
C₃H₄N₂S Boiling Pt: 117 °C/15 mm Melting Pt: 88…92 °C Density: 1.5 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00005325 CAS Number: 96-50-4 EINECS: 202-511-6 Merck Index: 14,479 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...