Guanine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-Amino-1H-purin-6(7H)-one
A12024-36
A12024-14
A12024-22
CAAAAA12024-36EA
443.98
CAD
CAAAAA12024-36
CAAAA12024-14
CAAAA12024-22
Guanine 98%
Guanine
MDL: MFCD00071533
Beilstein Registry No.: 9680
Purity: 98%
CAS No.: 73-40-5
Molecular Formula: C5H5N5O
Formula Weight: 151.13
Storage Temperature: 2°C to 8°C
Melting Point:
Formula:
C₅H₅N₅O MW: 151.13 g/mol Melting Pt: >300 °C Density: 1.67 g/cm³ (20 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00071533 CAS Number: 73-40-5 EINECS: 200-799-8 Merck Index: 14,4564 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...