(4-Carboxybutyl)triphenylphosphonium bromide 98%
Supplier: Thermo Scientific Chemicals
A12023-14
A12023-22
A12023-36
CAAAA12023-14EA
85.48
CAD
CAAAA12023-14
CAAAA12023-22
CAAAA12023-36
(4-Carboxybutyl)triphenylphosphonium bromide 98%
(4-Carboxybutyl)triphenylphosphonium bromide
MDL: MFCD00011906
Beilstein Registry No.: 3586477
Purity: 98%
CAS No.: 17814-85-6
Molecular Formula: C23H24BrO2P
Formula Weight: 443.33
Flash Point: 195°(383°F)
Melting Point: 205-209°
Beilstein Reference: 3586477
MDL No.: MFCD00011906
Formula:
C₂₃H₂₄BrO₂P Melting Pt: 205…209 °C Flash Pt: 195 °C(383 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00011906 CAS Number: 17814-85-6 EINECS: 241-782-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...